missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorophenol, 98+%, Thermo Scientific Chemicals
28.13 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic Can
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 98.0 |
CAS Max % | 100.0 |
Melting Point | 8.0°C |
Density | 1.2620g/mL |
Boiling Point | 173.0°C to 175.0°C |
Flash Point | 63°C |
Assay Percent Range | 98% min. (GC) |
Packaging | Glass bottle |
Linear Formula | ClC6H4OH |
Refractive Index | 1.5580 to 1.5600 |
Show More |
Safety and Handling
Product Identifier
- Sodium nitrite
- Danger
- Oxidising solid Category 3
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- ACUTE AQUATIC Acute 1
- H272-May intensify fire; oxidiser.
- H301-Toxic if swallowed.
- H319-Causes serious eye irritation.
- H400-Very toxic to aquatic life.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P273-Avoid release to the environment.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'