missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenethyl bromide, 95%, Thermo Scientific Chemicals

Catalog Number 11924541
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
16793-91-2
C8H8BrCl
219.51
AECBVDLERUETKG-UHFFFAOYSA-N
2756963
1-(2-bromoethyl)-2-chlorobenzene
C1=CC=C(C(=C1)CCBr)Cl
This item is not returnable. View return policy
16793-91-2
C8H8BrCl
219.51
AECBVDLERUETKG-UHFFFAOYSA-N
2756963
1-(2-bromoethyl)-2-chlorobenzene
C1=CC=C(C(=C1)CCBr)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
Color Colorless to Yellow
CAS Max % 100.0
Density 1.4569g/mL
Boiling Point 110°C to 112°C (10.0 Torr)
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range 94% min. (GC)
Specific Gravity 1.4569
Synonym 2-chlorophenethyl bromide, 1-2-bromoethyl-2-chlorobenzene, 2-2-chlorophenyl ethylbromide, 1-2-bromo-ethyl-2-chloro-benzene, 1-2-bromo-ethyl-2-chlorobenzene, benzene,1-2-bromoethyl-2-chloro, benzene, 1-2-bromoethyl-2-chloro, 1-2-bromoethyl-2-chloro-benzene, 2-chlorophenethylbromide, acmc-1cfee
Formula Weight 219.51
Percent Purity 95%
Chemical Name or Material 2-Chlorophenethyl bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Chlorophenethyl bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'