missing translation for 'onlineSavingsMsg'
Learn More

2-Chloronicotinamide, 98%, Thermo Scientific™

Catalog Number p-89641
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
10366-35-5
C6H5ClN2O
156.57
MFCD00006237
ZQZAHPFFZWEUCL-UHFFFAOYSA-N
2-chloronicotinamide, 3-pyridinecarboxamide, 2-chloro, chloronicotinamide, 2-chloro-nicotinamide, acmc-1bvtu, nicotinamide, 2-chloro, 2-chloronicotinic acid amide, ksc492e7f, 2-chloro-3-pyridinecarboxamide
82588
2-chloropyridine-3-carboxamide
C1=CC(=C(N=C1)Cl)C(=O)N
This item is not returnable. View return policy
10366-35-5
C6H5ClN2O
156.57
MFCD00006237
ZQZAHPFFZWEUCL-UHFFFAOYSA-N
2-chloronicotinamide, 3-pyridinecarboxamide, 2-chloro, chloronicotinamide, 2-chloro-nicotinamide, acmc-1bvtu, nicotinamide, 2-chloro, 2-chloronicotinic acid amide, ksc492e7f, 2-chloro-3-pyridinecarboxamide
82588
2-chloropyridine-3-carboxamide
C1=CC(=C(N=C1)Cl)C(=O)N
CAS Min % 97.5
CAS Max % 100.0
Melting Point 162°C to 166°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula Cl(C5H3N)CONH2
Quantity 5 g
Beilstein 22, II, 36
Formula Weight 156.57
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Chloronicotinamide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloronicotinamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'