missing translation for 'onlineSavingsMsg'
Learn More

2-Chloromethylbenzimidazole, 95%, Thermo Scientific Chemicals

Catalog Number p-88612
23.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4857-04-9
C8H7ClN2
166.61
MFCD00005602
SPMLMLQATWNZEE-UHFFFAOYSA-N
2-(chloromethyl)-1H-1,3-benzodiazole
ClCC1=NC2=CC=CC=C2N1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4857-04-9
C8H7ClN2
166.61
MFCD00005602
SPMLMLQATWNZEE-UHFFFAOYSA-N
2-(chloromethyl)-1H-1,3-benzodiazole
ClCC1=NC2=CC=CC=C2N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 145°C to 150°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 166.61
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Chloromethylbenzimidazole, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloromethylbenzimidazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'