missing translation for 'onlineSavingsMsg'
Learn More

2-Chloromethyl-1,4-benzodioxane, 97%, Thermo Scientific™

Catalog Number p-7044028
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
2164-33-2
C9H9ClO2
184.619
MFCD00014666
AYPKYQSHFKQVDL-UHFFFAOYSA-N
2-chloromethyl-2,3-dihydro-1,4-benzodioxine, 2-chloromethyl-1,4-benzodioxane, 2-chloromethyl-2,3-dihydrobenzo b 1,4 dioxine, 2-chloromethyl-2,3-dihydro-1,4-benzodioxin, 2-chloromethyl-2,3-dihydro-benzo 1,4 dioxine, 2-chloromethyl-2h,3h-benzo e 1,4-dioxane, acmc-1cfd2, 2-chloromethyl-benzodioxane, 2-chloromethyl-1,4-benzodioxan
95320
3-(chloromethyl)-2,3-dihydro-1,4-benzodioxine
C1C(OC2=CC=CC=C2O1)CCl
This item is not returnable. View return policy
2164-33-2
C9H9ClO2
184.619
MFCD00014666
AYPKYQSHFKQVDL-UHFFFAOYSA-N
2-chloromethyl-2,3-dihydro-1,4-benzodioxine, 2-chloromethyl-1,4-benzodioxane, 2-chloromethyl-2,3-dihydrobenzo b 1,4 dioxine, 2-chloromethyl-2,3-dihydro-1,4-benzodioxin, 2-chloromethyl-2,3-dihydro-benzo 1,4 dioxine, 2-chloromethyl-2h,3h-benzo e 1,4-dioxane, acmc-1cfd2, 2-chloromethyl-benzodioxane, 2-chloromethyl-1,4-benzodioxan
95320
3-(chloromethyl)-2,3-dihydro-1,4-benzodioxine
C1C(OC2=CC=CC=C2O1)CCl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 96°C (0.4 mmHg)
Refractive Index 1.554
Quantity 5 g
Beilstein 150153
Formula Weight 184.62
Percent Purity 97%
Chemical Name or Material 2-Chloromethyl-1,4-benzodioxane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'