missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-2-Chloroisodurene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 34°C to 38°C |
Color | White |
Boiling Point | 114°C to 115°C (10.0 mmHg) |
Flash Point | 106°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | (CH3)3C6H2CH2Cl |
Show More |
Safety and Handling
Product Identifier
- alpha-2-Chloroisodurene
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P390-Absorb spillage to prevent material damage.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'