missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog Number p-7045689
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
6099-88-3
C3H4ClNS
121.582
MFCD00041125
ZUWFBQUHBOUPFK-UHFFFAOYSA-N
2-chloroethyl isothiocyanate, 2-chloroethylisothiocyanate, ethane, 1-chloro-2-isothiocyanato, sgxdlhbabrkth@, chloroethylisothiocyanate, acmc-1b2t4, beta-chloroethylisothiocyanate, beta-chloroethyl isothiocyanate, 1-chloro-2-isothiocyanato-ethane, #
138661
1-chloro-2-isothiocyanatoethane
C(CCl)N=C=S
This item is not returnable. View return policy
6099-88-3
C3H4ClNS
121.582
MFCD00041125
ZUWFBQUHBOUPFK-UHFFFAOYSA-N
2-chloroethyl isothiocyanate, 2-chloroethylisothiocyanate, ethane, 1-chloro-2-isothiocyanato, sgxdlhbabrkth@, chloroethylisothiocyanate, acmc-1b2t4, beta-chloroethylisothiocyanate, beta-chloroethyl isothiocyanate, 1-chloro-2-isothiocyanato-ethane, #
138661
1-chloro-2-isothiocyanatoethane
C(CCl)N=C=S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.265
Boiling Point 79°C to 80°C (13 mmHg)
Flash Point 96°C (205°F)
Refractive Index 1.556
Quantity 1 g
UN Number UN2922
Beilstein 605585
Sensitivity Moisture sensitive
Formula Weight 121.59
Percent Purity 97%
Chemical Name or Material 2-Chloroethyl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'