missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethanol, 99%, Thermo Scientific Chemicals

Catalog Number 15491457
Click to view available options
Quantity:
50 mL
250 mL
1 L
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
107-07-3
C2H5ClO
80.511
MFCD00002829
SZIFAVKTNFCBPC-UHFFFAOYSA-N
ethylene chlorohydrin, ethanol, 2-chloro, glycol chlorohydrin, 2-chloroethyl alcohol, chloroethanol, glycol monochlorohydrin, 2-monochloroethanol, ethylchlorohydrin, 2-chlorethanol, 2-chloro-1-ethanol
34
CHEBI:28200
2-chloroethanol
C(CCl)O
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
107-07-3
C2H5ClO
80.511
MFCD00002829
SZIFAVKTNFCBPC-UHFFFAOYSA-N
ethylene chlorohydrin, ethanol, 2-chloro, glycol chlorohydrin, 2-chloroethyl alcohol, chloroethanol, glycol monochlorohydrin, 2-monochloroethanol, ethylchlorohydrin, 2-chlorethanol, 2-chloro-1-ethanol
34
CHEBI:28200
2-chloroethanol
C(CCl)O

2-Chloroethanol is used in the preparation of several dyes by the alkylation of aniline derivatives with chloroethanol. It is a building block in the production of pharmaceuticals and plasticizers. Also used for manufacture of thiodiglycol. It is a solvent for cellulose acetate and ethyl cellulose, textile printing dyes, in dewaxing, refining of rosin, extraction of pine lignin and the cleaning of machines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point °C to 63°C
Density 1.202
Boiling Point 129°C
Flash Point 55°C (131°F)
Linear Formula CH2CH2OH
Refractive Index 1.441
Quantity 250 mL
UN Number UN1135
Beilstein 878139
Merck Index 14,3793
Solubility Information Fully miscible with water.
Formula Weight 80.51
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Chloroethanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'