missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 2-Chlorocyclopentanone, 98%, stab. with potassium carbonate, Thermo Scientific™
Click to view available options
Quantity:
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Chlorocyclopentanone is used as pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
2-Chlorocyclopentanone is used as pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Density | 1.185 |
Boiling Point | 72°C to 74°C (12 mmHg) |
Flash Point | 77°C (171°F) |
Refractive Index | 1.4745 |
Quantity | 5 g |
Beilstein | 1098899 |
Solubility Information | Soluble in water. |
Formula Weight | 118.56 |
Percent Purity | 98% |
Chemical Name or Material | 2-Chlorocyclopentanone, stabilized with potassium carbonate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'