missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chlorobutyryl chloride, tech. 85%

Catalog Number 11307636
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
7623-11-2
MFCD00041409
This item is not returnable. View return policy
7623-11-2
MFCD00041409
Applications
2-Chlorobutyryl chloride is used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY
Density 1.236
Boiling Point 138°C to 140°C
Molecular Formula C3H4Cl2O
Refractive Index 1.451
Quantity 25 g
UN Number UN3265
Beilstein 741947
Sensitivity Moisture sensitive
Synonym 2-chloropropionyl chloride, propanoyl chloride, 2-chloro, alpha-chloropropionyl chloride, 2-chloropropionic acid chloride, 2-chloropropionylchloride, .alpha.-chloropropionyl chloride, 2-chloropropanyl chloride, 1,2dichloro-1-propanone, 2-chloro-propionylchloride, 1,2-dichloro-1-propanone
Solubility Information Reacts with water.
InChI Key JEQDSBVHLKBEIZ-UHFFFAOYNA-N
SMILES CC(C(=O)Cl)Cl
IUPAC Name 2-chloropropanoyl chloride
Molecular Weight (g/mol) 126.96
PubChem CID 111019
Formula Weight 141
Percent Purity 85%
Grade Technical
Chemical Name or Material 2-Chlorobutyryl chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'