missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl mercaptan, 98%, Thermo Scientific™

Catalog Number p-3211662
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
39718-00-8
C7H7ClS
158.65
MFCD00004868
WWFIIZLHSNBNTC-UHFFFAOYSA-N
2-chlorobenzenemethanethiol, 2-chlorophenyl methanethiol, 2-chlorobenzyl mercaptan, 2-chlorobenzylmercaptan, 2-chloro benzyl mercaptan, benzenemethanethiol, 2-chloro, o-chlorobenzylmercaptan, pubchem6836, 2-chlorophenyl-methanethiol, d0p3xw
580759
(2-chlorophenyl)methanethiol
C1=CC=C(C(=C1)CS)Cl
This item is not returnable. View return policy
39718-00-8
C7H7ClS
158.65
MFCD00004868
WWFIIZLHSNBNTC-UHFFFAOYSA-N
2-chlorobenzenemethanethiol, 2-chlorophenyl methanethiol, 2-chlorobenzyl mercaptan, 2-chlorobenzylmercaptan, 2-chloro benzyl mercaptan, benzenemethanethiol, 2-chloro, o-chlorobenzylmercaptan, pubchem6836, 2-chlorophenyl-methanethiol, d0p3xw
580759
(2-chlorophenyl)methanethiol
C1=CC=C(C(=C1)CS)Cl
CAS Min % 97.5
Color Colorless to Yellow
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula ClC6H4CH2SH
Formula Weight 158.65
Percent Purity 98%
Density 1.2200g/mL
Boiling Point 120°C to 121°C (25.0 mmHg)
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5890 to 1.591
Specific Gravity 1.22
Chemical Name or Material 2-Chlorobenzyl mercaptan
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chlorobenzyl mercaptan
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'