missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl mercaptan, 97%

Product Code. 11402508
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11402508

Brand: Thermo Scientific Alfa Aesar A15826.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 39718-00-8 | C7H7ClS | 158.643 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39718-00-8
Molecular Formula C7H7ClS
Molecular Weight (g/mol) 158.643
MDL Number MFCD00004868
InChI Key WWFIIZLHSNBNTC-UHFFFAOYSA-N
Synonym 2-chlorobenzenemethanethiol, 2-chlorophenyl methanethiol, 2-chlorobenzyl mercaptan, 2-chlorobenzylmercaptan, 2-chloro benzyl mercaptan, benzenemethanethiol, 2-chloro, o-chlorobenzylmercaptan, pubchem6836, 2-chlorophenyl-methanethiol, d0p3xw
PubChem CID 580759
IUPAC Name (2-chlorophenyl)methanethiol
SMILES C1=CC=C(C(=C1)CS)Cl

Specifications

Density 1.22
Boiling Point 120°C to 122°C (25 mmHg)
Quantity 25 g
Refractive Index 1.591
UN Number UN2810
Beilstein 2042183
Formula Weight 158.65
Percent Purity 97%
Chemical Name or Material 2-Chlorobenzyl mercaptan

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.