missing translation for 'onlineSavingsMsg'
Learn More

2-chlorobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog Number p-3777845
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
611-17-6
C7H7BrCl
205.49
MFCD00000566
PURSZYWBIQIANP-UHFFFAOYSA-N
2-chlorobenzyl bromide, 1-bromomethyl-2-chlorobenzene, o-chlorobenzyl bromide, 2-chlorobenzylbromide, benzene, 1-bromomethyl-2-chloro, alpha-bromo-2-chlorotoluene, unii-cev23h8uv4, 2chlorobenzyl bromide, alpha-bromo-o-chlorotoluene, cev23h8uv4
11905
1-(bromomethyl)-2-chlorobenzene
C1=CC=C(C(=C1)CBr)Cl
This item is not returnable. View return policy
611-17-6
C7H7BrCl
205.49
MFCD00000566
PURSZYWBIQIANP-UHFFFAOYSA-N
2-chlorobenzyl bromide, 1-bromomethyl-2-chlorobenzene, o-chlorobenzyl bromide, 2-chlorobenzylbromide, benzene, 1-bromomethyl-2-chloro, alpha-bromo-2-chlorotoluene, unii-cev23h8uv4, 2chlorobenzyl bromide, alpha-bromo-o-chlorotoluene, cev23h8uv4
11905
1-(bromomethyl)-2-chlorobenzene
C1=CC=C(C(=C1)CBr)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.5860g/mL
Boiling Point 103°C to 104°C (10.0 mmHg)
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula ClC6H4CH2Br
Refractive Index 1.5910 to 1.593
Quantity 5 g
Beilstein 05, 155
Specific Gravity 1.586
Formula Weight 205.49
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-chlorobenzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chlorobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'