missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog Number p-85118
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
17849-38-6
C7H7ClO
142.58
MFCD00004604
MBYQPPXEXWRMQC-UHFFFAOYSA-N
2-chlorobenzyl alcohol, 2-chlorophenyl methanol, benzenemethanol, 2-chloro, 2-chlorobenzenemethanol, o-chlorobenzyl alcohol, benzyl alcohol, o-chloro, 2-chloro-phenyl-methanol, 2-chlorophenyl methan-1-ol, benzenemethanol, ar-chloro, 2-chlorobenzylalcohol
28810
(2-chlorophenyl)methanol
C1=CC=C(C(=C1)CO)Cl
This item is not returnable. View return policy
17849-38-6
C7H7ClO
142.58
MFCD00004604
MBYQPPXEXWRMQC-UHFFFAOYSA-N
2-chlorobenzyl alcohol, 2-chlorophenyl methanol, benzenemethanol, 2-chloro, 2-chlorobenzenemethanol, o-chlorobenzyl alcohol, benzyl alcohol, o-chloro, 2-chloro-phenyl-methanol, 2-chlorophenyl methan-1-ol, benzenemethanol, ar-chloro, 2-chlorobenzylalcohol
28810
(2-chlorophenyl)methanol
C1=CC=C(C(=C1)CO)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 69°C to 72°C
Color White
Boiling Point 227°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4CH2OH
Quantity 10 g
Beilstein 06, 444
Solubility Information Solubility in water: limited soluble in water. Other solubilities: 1g/10mL in methanol
Formula Weight 142.58
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Chlorobenzyl alcohol, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'