missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzothiazole, 99%, Thermo Scientific Chemicals

Catalog Number p-2698000
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
615-20-3
C7H4ClNS
169.63
BSQLQMLFTHJVKS-UHFFFAOYSA-N
2-chlorobenzothiazole, benzothiazole, 2-chloro, 2-chlorobenzo d thiazole, 2-benzothiazolyl chloride, usaf ek-2784, 2-chloro-benzothiazole, benzothiazole, chloro, unii-fn1t1y0p5p, fn1t1y0p5p, 2-chlorobenzthiazole
11987
2-chloro-1,3-benzothiazole
C1=CC=C2C(=C1)N=C(S2)Cl
This item is not returnable. View return policy
615-20-3
C7H4ClNS
169.63
BSQLQMLFTHJVKS-UHFFFAOYSA-N
2-chlorobenzothiazole, benzothiazole, 2-chloro, 2-chlorobenzo d thiazole, 2-benzothiazolyl chloride, usaf ek-2784, 2-chloro-benzothiazole, benzothiazole, chloro, unii-fn1t1y0p5p, fn1t1y0p5p, 2-chlorobenzthiazole
11987
2-chloro-1,3-benzothiazole
C1=CC=C2C(=C1)N=C(S2)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Brown-Yellow to Yellow
Density 1.3700g/mL
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.6367 to 1.6387
Quantity 5 g
Beilstein 27, 44
Specific Gravity 1.37
Solubility Information Solubility in water: insoluble
Formula Weight 169.63
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Chlorobenzothiazole
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chlorobenzothiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'