missing translation for 'onlineSavingsMsg'
Learn More

2'-Chloroacetophenone, 97%, Thermo Scientific Chemicals

Catalog Number p-84891
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2142-68-9
C8H7ClO
154.6
MFCD00000560
ZDOYHCIRUPHUHN-UHFFFAOYSA-N
2'-chloroacetophenone, 1-2-chlorophenyl ethanone, o-chloroacetophenone, 1-2-chlorophenyl ethan-1-one, ethanone, 1-2-chlorophenyl, acetophenone, 2'-chloro, o-chloroacetophonone, 1-2-chloro-phenyl-ethanone, ethanone, 1-chlorophenyl, 1-acetyl-2-chlorobenzene
72864
1-(2-chlorophenyl)ethanone
CC(=O)C1=CC=CC=C1Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2142-68-9
C8H7ClO
154.6
MFCD00000560
ZDOYHCIRUPHUHN-UHFFFAOYSA-N
2'-chloroacetophenone, 1-2-chlorophenyl ethanone, o-chloroacetophenone, 1-2-chlorophenyl ethan-1-one, ethanone, 1-2-chlorophenyl, acetophenone, 2'-chloro, o-chloroacetophonone, 1-2-chloro-phenyl-ethanone, ethanone, 1-chlorophenyl, 1-acetyl-2-chlorobenzene
72864
1-(2-chlorophenyl)ethanone
CC(=O)C1=CC=CC=C1Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Amber to Yellow
Density 1.1880g/mL
Boiling Point 228.0°C to 229.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4COCH3
Refractive Index 1.5420 to 1.5450
Quantity 25 g
Beilstein 07, I, 151
Specific Gravity 1.188
Formula Weight 154.6
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2'-Chloroacetophenone, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'