missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroacetamide, 98%, Thermo Scientific Chemicals
13.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
13.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "22914" to get your promotional price.
13.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
500 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 116.0°C to 120.0°C |
Color | White |
Density | 0.8400g/mL |
Boiling Point | 224.5°C |
Flash Point | 170°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- 2-Chloroacetamide
- Danger
- Acute toxicity Category 3
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H361f-Suspected of damaging fertility.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'