Learn More
Alfa Aesar™ 2-Chloro-N-phenylbenzamide, 97%
Brand: Alfa Aesar™ H59623.03
Code : NEW
Additional Details : CAS Number : 6833-13-2 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
2-Chloro-N-phenylbenzamide | |
6833-13-2 | |
231.68 | |
MFCD00016336 |
Specifications
2-Chloro-N-phenylbenzamide | |
6833-13-2 | |
97% | |
MFCD00016336 |
1g | |
Solid | |
231.68 |