missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-N,N-di-n-propylnicotinamide, 97%

Brand:  Alfa Aesar™ H57061.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15493608

  • £89.68 / 1g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-N,N-di-n-propylnicotinamide
240.73
MFCD01343305
2-Chloro-N,N-di-n-propylpyridine-3-carboxamide

Specifications

2-Chloro-N,N-di-n-propylnicotinamide
1g
97%
MFCD01343305
Pale yellow to brown
Liquid
240.73
2-Chloro-N,N-di-n-propylpyridine-3-carboxamide
Videos
SDS