missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-N-(5,6,7,8-tetrahydro-1-naphthyl)-4-(trifluoromethyl)benzamide, 95%

Brand:  Alfa Aesar™ H65656.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429816

  • £79.71 / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-N-(5,6,7,8-tetrahydro-1-naphthyl)-4-(trifluoromethyl)benzamide
353.77
MFCD27975800

Specifications

2-Chloro-N-(5,6,7,8-tetrahydro-1-naphthyl)-4-(trifluoromethyl)benzamide
Solid
353.77
1g
95%
MFCD27975800
Videos
SDS