missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-N-(4-fluoro-2-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59315.03

Code : NEW

Additional Details : CAS Number : 895665-46-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15465588

  • £358.49 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-N-(4-fluoro-2-methylphenyl)benzamide
895665-46-0
263.7
MFCD07264147

Specifications

2-Chloro-N-(4-fluoro-2-methylphenyl)benzamide
1g
97%
MFCD07264147
895665-46-0
Solid
263.7
Videos
SDS