Alfa Aesar™ 2-Chloro-N-(4-ethoxyphenyl)benzamide, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H59553.MD

250MG 2-Chloro-N-(4-ethoxyphenyl)benzamide, 97% 250mg

UNSPSC: 12352106

Code: NEW

Additional Details:
Additional Details: CAS Number: 196803-92-6 Weight: 0.00025kg


Quantity 250mg

Product Code. 15405918

Quantity Price
1 £ 111.72 / 250mg
Estimated Shipment date
from Supplier 10-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2-Chloro-N-(4-ethoxyphenyl)benzamide
CAS 196803-92-6
Formula Weight 275.73
MDL Number MFCD00157705

Specification

Formula Weight 275.73
Chemical Name or Material 2-Chloro-N-(4-ethoxyphenyl)benzamide
Quantity 250mg
Physical Form Solid
Assay 97%
MDL Number MFCD00157705
CAS 196803-92-6