missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-N-(3-fluoro-5-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59546.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15425908

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-07-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-N-(3-fluoro-5-methylphenyl)benzamide
263.7
MFCD22683076

Specifications

2-Chloro-N-(3-fluoro-5-methylphenyl)benzamide
White
97%
MFCD22683076
1g
Solid
263.7
Videos
SDS