missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-N-(3-fluoro-2-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59303.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15405578

  • £358.49 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-N-(3-fluoro-2-methylphenyl)benzamide
263.7
MFCD22683054

Specifications

2-Chloro-N-(3-fluoro-2-methylphenyl)benzamide
Off-white to pale yellow
97%
MFCD22683054
1g
Solid
263.7
Videos
SDS