Alfa Aesar™ 2-Chloro-N-(3-chloro-4-methoxyphenyl)benzamide, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H59004.03

UNSPSC: 12352106

Code: NEW

Additional Details:
Additional Details: CAS Number: 443903-11-5 Weight: 0.00100kg


Quantity 1g

Product Code. 15404716

Quantity Price
1 £ 358.49 / 1g
Estimated Shipment date
from Supplier 10-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2-Chloro-N-(3-chloro-4-methoxyphenyl)benzamide
CAS 443903-11-5
Formula Weight 296.15
MDL Number MFCD01608894

Specification

Formula Weight 296.15
Quantity 1g
MDL Number MFCD01608894
Assay 97%
CAS 443903-11-5
Physical Form Solid
Chemical Name or Material 2-Chloro-N-(3-chloro-4-methoxyphenyl)benzamide
Documentation

Technical documents