missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-N-(3,5-difluorophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59458.03

Code : NEW

Additional Details : CAS Number : 881764-20-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15475788

  • £358.49 / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-N-(3,5-difluorophenyl)benzamide
881764-20-1
267.66
MFCD07081996

Specifications

2-Chloro-N-(3,5-difluorophenyl)benzamide
1g
97%
MFCD07081996
881764-20-1
Solid
267.66
Videos
SDS