missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-N-(3,5-difluorophenyl)-4-nitrobenzamide, 97%

Brand:  Alfa Aesar™ H59962.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15485376

  • £358.49 / 1g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-N-(3,5-difluorophenyl)-4-nitrobenzamide
312.66
MFCD06634000

Specifications

2-Chloro-N-(3,5-difluorophenyl)-4-nitrobenzamide
Solid
312.66
1g
97%
MFCD06634000
Videos
SDS