missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-N-(2-fluoro-5-methylphenyl)-4-nitrobenzamide, 97%

Brand:  Alfa Aesar™ H59642.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415156

  • £358.49 / 1g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-N-(2-fluoro-5-methylphenyl)-4-nitrobenzamide
308.69
MFCD15344955

Specifications

2-Chloro-N-(2-fluoro-5-methylphenyl)-4-nitrobenzamide
Solid
308.69
1g
97%
MFCD15344955
Videos
SDS