missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-N-(2-fluoro-4-methylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59631.03

Code : NEW

Additional Details : CAS Number : 878966-10-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15416018

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-N-(2-fluoro-4-methylphenyl)benzamide
878966-10-0
263.7
MFCD07848474

Specifications

2-Chloro-N-(2-fluoro-4-methylphenyl)benzamide
1g
97%
MFCD07848474
878966-10-0
Solid
263.7
Videos
SDS