missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-N-(2-fluoro-4-iodophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59304.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415578

  • £358.49 / 1g


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-N-(2-fluoro-4-iodophenyl)benzamide
375.57
MFCD22683055

Specifications

2-Chloro-N-(2-fluoro-4-iodophenyl)benzamide
Light Sensitive
97%
MFCD22683055
1g
Solid
375.57
Videos
SDS