missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-N-(2-ethoxyphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59019.03

Code : NEW

Additional Details : CAS Number : 196803-91-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15475188

  • £358.49 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-N-(2-ethoxyphenyl)benzamide
196803-91-5
275.73
MFCD00090977

Specifications

2-Chloro-N-(2-ethoxyphenyl)benzamide
1g
Viscous Liquid
275.73
196803-91-5
Brown
97%
MFCD00090977
Videos
SDS