missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-N-(2-chloro-4-iodophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59566.03

Code : NEW

Additional Details : CAS Number : 424815-36-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15405938

  • £358.49 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-N-(2-chloro-4-iodophenyl)benzamide
424815-36-1
392.02
MFCD02216270

Specifications

2-Chloro-N-(2-chloro-4-iodophenyl)benzamide
1g
Solid
392.02
424815-36-1
Light Sensitive
97%
MFCD02216270
Videos
SDS