missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-N-(2,5-difluorophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59149.03

Code : NEW

Additional Details : CAS Number : 903302-26-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15495368

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-N-(2,5-difluorophenyl)benzamide
903302-26-1
267.66
MFCD04680213

Specifications

2-Chloro-N-(2,5-difluorophenyl)benzamide
1g
97%
MFCD04680213
903302-26-1
Solid
267.66
Videos
SDS