Alfa Aesar™ 2-Chloro-N-(2,4-dimethoxyphenyl)benzamide, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H59226.MD

UNSPSC: 12352106

Code: NEW

Additional Details:
Additional Details: CAS Number: 196938-29-1 Weight: 0.00025kg


Quantity 250mg

Product Code. 15464896

Quantity Price
1 £ 111.72 / 250mg
Estimated Shipment date
from Supplier 13-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2-Chloro-N-(2,4-dimethoxyphenyl)benzamide
CAS 196938-29-1
Formula Weight 291.73
MDL Number MFCD00032669

Specification

Formula Weight 291.73
Chemical Name or Material 2-Chloro-N-(2,4-dimethoxyphenyl)benzamide
CAS 196938-29-1
Physical Form Solid
Assay 97%
Quantity 250mg
MDL Number MFCD00032669