missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-N-(2,4-difluorophenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59975.03

Code : NEW

Additional Details : CAS Number : 349125-64-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15426438

  • £358.49 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-N-(2,4-difluorophenyl)benzamide
349125-64-0
267.66
MFCD00522770

Specifications

2-Chloro-N-(2,4-difluorophenyl)benzamide
1g
97%
MFCD00522770
349125-64-0
Solid
267.66
Videos
SDS