missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-N-(2,4-dichlorophenyl)-4-nitrobenzamide, 97%

Brand:  Alfa Aesar™ H59441.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15445026

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-N-(2,4-dichlorophenyl)-4-nitrobenzamide
345.57
MFCD00751429

Specifications

2-Chloro-N-(2,4-dichlorophenyl)-4-nitrobenzamide
Solid
345.57
1g
97%
MFCD00751429
Videos
SDS