missing translation for 'onlineSavingsMsg'
Learn More

(+)-2-Chloro-L-phenylglycine, 95%, Thermo Scientific™

Catalog Number 11432140
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
141315-50-6
C8H8ClNO2
185.607
MFCD03426346
LMIZLNPFTRQPSF-ZETCQYMHSA-N
s-2-amino-2-2-chlorophenyl acetic acid, l-2-chlorophenylglycine, l-2-chlorophenyl glycine, l-+-2-chlorophenylglycine, s-+-2-chlorophenylglycine, 2s-2-amino-2-2-chlorophenyl acetic acid, s-alpha-amino-2-chlorophenyl acetic acid, s-amino-2-chloro-phenyl-acetic acid, s-+-a-amino-2-chlorophenyl acetic acid, h-phg 2-cl-oh
1501937
(2S)-2-amino-2-(2-chlorophenyl)acetic acid
C1=CC=C(C(=C1)C(C(=O)O)N)Cl
This item is not returnable. View return policy
141315-50-6
C8H8ClNO2
185.607
MFCD03426346
LMIZLNPFTRQPSF-ZETCQYMHSA-N
s-2-amino-2-2-chlorophenyl acetic acid, l-2-chlorophenylglycine, l-2-chlorophenyl glycine, l-+-2-chlorophenylglycine, s-+-2-chlorophenylglycine, 2s-2-amino-2-2-chlorophenyl acetic acid, s-alpha-amino-2-chlorophenyl acetic acid, s-amino-2-chloro-phenyl-acetic acid, s-+-a-amino-2-chlorophenyl acetic acid, h-phg 2-cl-oh
1501937
(2S)-2-amino-2-(2-chlorophenyl)acetic acid
C1=CC=C(C(=C1)C(C(=O)O)N)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 182°C to 187°C
Formula Weight 185.61
Percent Purity 95%
Quantity 1 g
Optical Rotation Not completely soluble in Chloroform
Chemical Name or Material (+)-2-Chloro-L-phenylglycine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'