missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-7-nitroquinoxaline, 95%

Brand:  Alfa Aesar™ H64623.03

Code : NEW

Additional Details : CAS Number : 55686-94-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15421909

  • £119.78 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-7-nitroquinoxaline
209.59
172800
55686-94-7
MFCD00033669

Specifications

2-Chloro-7-nitroquinoxaline
1g
Solid
209.59
172800
185° to 190°C
55686-94-7
95%
MFCD00033669
Videos
SDS