missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylphenyl isothiocyanate, 95+%

Brand:  Maybridge SB01714EB

Code : 31

Additional Details : CAS Number : 19241-34-0 Weight : 0.07500kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III

Quantity 25g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10768465

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2-Chloro-6-methylphenyl isothiocyanate
95%
183.66
95%
C8H6ClNS

Specifications

2-Chloro-6-methylphenyl isothiocyanate
Amber Glass Bottle
95%
C8H6ClNS
19241-34-0
95%
25g
183.66
Videos
SDS