missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylisonicotinic acid, 95+%, Thermo Scientific™

Catalog Number p-4510365
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
25462-85-5
C7H6ClNO2
171.58
ZGZMEKHQIZSZOH-UHFFFAOYSA-N
2-chloro-6-methylisonicotinic acid, 2-chloro-6-picoline-4-carboxylic acid, 4-pyridinecarboxylic acid, 2-chloro-6-methyl, 2-methyl-6-chloropyridine-4-carboxylic acid, 2-chloro-6-methyl-pyridine-4-carboxylic acid, 2-chloro-6-methyl-4-pyridinecarboxylic acid, 2-chloro-6-methyl-isonicotinic acid, 2-chloro-6-methyl-4-pyridinecarboxylicacid, 2-chloro-6-methylisonicotinicacid, pubchem17665
141209
2-chloro-6-methylpyridine-4-carboxylic acid
CC1=NC(=CC(=C1)C(=O)O)Cl
This item is not returnable. View return policy
25462-85-5
C7H6ClNO2
171.58
ZGZMEKHQIZSZOH-UHFFFAOYSA-N
2-chloro-6-methylisonicotinic acid, 2-chloro-6-picoline-4-carboxylic acid, 4-pyridinecarboxylic acid, 2-chloro-6-methyl, 2-methyl-6-chloropyridine-4-carboxylic acid, 2-chloro-6-methyl-pyridine-4-carboxylic acid, 2-chloro-6-methyl-4-pyridinecarboxylic acid, 2-chloro-6-methyl-isonicotinic acid, 2-chloro-6-methyl-4-pyridinecarboxylicacid, 2-chloro-6-methylisonicotinicacid, pubchem17665
141209
2-chloro-6-methylpyridine-4-carboxylic acid
CC1=NC(=CC(=C1)C(=O)O)Cl
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 171.58
Percent Purity 95%
Chemical Name or Material 2-Chloro-6-methylisonicotinic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'