missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-chloro-6-methyl-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
10 g
50 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 5 g |
Formula Weight | 195.57 |
Percent Purity | 97% |
Chemical Name or Material | 2-Chloro-6-methyl-4-(trifluoromethyl)pyridine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'