missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-6-methoxyisonicotinic acid, 97%, Thermo Scientific™

Catalog Number p-2698125
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
15855-06-8
C7H6ClNO3
187.579
MFCD00173928
PJQBTHQTVJMCFX-UHFFFAOYSA-N
2-chloro-6-methoxyisonicotinic acid, 2-chloro-6-methoxy-isonicotinic acid, 4-pyridinecarboxylic acid, 2-chloro-6-methoxy, 2-chloro-6-methoxy-4-pyridinecarboxylic acid, 2-chloro-6-methoxyisonicotinicacid, acmc-1bs54, ksc173i0h, 2-chloro-6-methoxypyridine-4-carboxylicacid
2782124
2-chloro-6-methoxypyridine-4-carboxylic acid
COC1=NC(=CC(=C1)C(=O)O)Cl
This item is not returnable. View return policy
15855-06-8
C7H6ClNO3
187.579
MFCD00173928
PJQBTHQTVJMCFX-UHFFFAOYSA-N
2-chloro-6-methoxyisonicotinic acid, 2-chloro-6-methoxy-isonicotinic acid, 4-pyridinecarboxylic acid, 2-chloro-6-methoxy, 2-chloro-6-methoxy-4-pyridinecarboxylic acid, 2-chloro-6-methoxyisonicotinicacid, acmc-1bs54, ksc173i0h, 2-chloro-6-methoxypyridine-4-carboxylicacid
2782124
2-chloro-6-methoxypyridine-4-carboxylic acid
COC1=NC(=CC(=C1)C(=O)O)Cl
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 187.58
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-methoxyisonicotinic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'