missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-6-fluorophenylboronic acid, 97%, Thermo Scientific™

Catalog Number p-4228509
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
313545-32-3
C6H5BClFO2
174.36
MFCD04039892
NXSZSZJWZVLHAY-UHFFFAOYSA-N
2-chloro-6-fluorophenyl boronic acid, 2-chloro-6-fluorobenzeneboronic acid, 2-chloro-6-fluorophenylboronicacid, boronic acid, 2-chloro-6-fluorophenyl, boronic acid,b-2-chloro-6-fluorophenyl, boronic acid, b-2-chloro-6-fluorophenyl, pubchem18883, acmc-1ad0v, 2-borono-1-chloro-3-fluorobenzene, 2-chloro-6-fluorobenzene boronic acid
3549395
(2-chloro-6-fluorophenyl)boronic acid
OB(O)C1=C(F)C=CC=C1Cl
This item is not returnable. View return policy
313545-32-3
C6H5BClFO2
174.36
MFCD04039892
NXSZSZJWZVLHAY-UHFFFAOYSA-N
2-chloro-6-fluorophenyl boronic acid, 2-chloro-6-fluorobenzeneboronic acid, 2-chloro-6-fluorophenylboronicacid, boronic acid, 2-chloro-6-fluorophenyl, boronic acid,b-2-chloro-6-fluorophenyl, boronic acid, b-2-chloro-6-fluorophenyl, pubchem18883, acmc-1ad0v, 2-borono-1-chloro-3-fluorobenzene, 2-chloro-6-fluorobenzene boronic acid
3549395
(2-chloro-6-fluorophenyl)boronic acid
OB(O)C1=C(F)C=CC=C1Cl
CAS Min % 96.0
CAS Max % 100.0
Melting Point 129.9°C to 132.8°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 174.37
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloro-6-fluorophenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-6-fluorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'