missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-6-fluorobenzylamine, 97%, ACROS Organics™

Brand:  Acros Organics 350631000

Code : 29

Additional Details : CAS Number : 15205-15-9 Weight : 0.15000kg Transport : UN number : 2735 Chem class : 1b Pack group : III


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10239373

  • £286.50 / 100g


Estimated Shipment
12-02-2018
Add to basket


Specifications

Chemical Identifiers

2-Chloro-6-fluorobenzylamine
96.0
≥96.0%
C7H7ClFN
159.59
15205-15-9
100.0
97%
ClC6H3(F)CH2NH2
MFCD00042458

Specifications

2-Chloro-6-fluorobenzylamine
Authentic
100g
96.0
100.0
Liquid
97%
ClC6H3(F)CH2NH2
MFCD00042458
1.2400g/mL
Clear colorless to light yellow
15205-15-9
Glass bottle
1.5340 to 1.5360 (20°C, 589nm)
≥96.0%
C7H7ClFN
159.59
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Chloro-6-fluorobenzylamine

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS