missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(pentafluoroethyl)pyridine, 96%, Thermo Scientific™

Catalog Number 15412809
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 118559-21-0
Color Yellow
Molecular Formula C7H3ClF5N
MDL Number MFCD17677154
Quantity 100 mg
Synonym 2-chloro-5-pentafluoroethylpyridine, 2-chloro-5-perfluoroethyl pyridine, 2-chloro-5-pentafluoroethyl pyridine, 2-chloro-5-pentafluoroethyl-pyridine, 2-chloro-5-1,1,2,2,2-pentafluoroethyl pyridine
InChI Key YRBHRYDSCMEMAY-UHFFFAOYSA-N
SMILES C1=CC(=NC=C1C(C(F)(F)F)(F)F)Cl
IUPAC Name 2-chloro-5-(1,1,2,2,2-pentafluoroethyl)pyridine
Molecular Weight (g/mol) 231.55
PubChem CID 14111645
Formula Weight 231.55
Percent Purity 96%
Physical Form Viscous Liquid
Chemical Name or Material 2-Chloro-5-(pentafluoroethyl)pyridine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'