missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrobenzophenone, 98%, Thermo Scientific Chemicals

Catalog Number p-7032955
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
34052-37-4
C13H8ClNO3
261.661
MFCD00007295
HRPHZUAPQWJPCZ-UHFFFAOYSA-N
2-chloro-5-nitrobenzophenone, 2-chloro-5-nitrophenyl phenyl methanone, 2-chloro-5-nitrophenyl-phenylmethanone, 2-chloro-5-nitrophenyl phenyl ketone, acmc-1aifx, methanone, 2-chloro-5-nitrophenyl phenyl, 5-nitro-2-chlorobenzophenone, 2-chloro-5-nitro-benzophenone, hrphzuapqwjpcz-uhfffaoysa
118591
(2-chloro-5-nitrophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)[N+](=O)[O-])Cl
This item is not returnable. View return policy
34052-37-4
C13H8ClNO3
261.661
MFCD00007295
HRPHZUAPQWJPCZ-UHFFFAOYSA-N
2-chloro-5-nitrobenzophenone, 2-chloro-5-nitrophenyl phenyl methanone, 2-chloro-5-nitrophenyl-phenylmethanone, 2-chloro-5-nitrophenyl phenyl ketone, acmc-1aifx, methanone, 2-chloro-5-nitrophenyl phenyl, 5-nitro-2-chlorobenzophenone, 2-chloro-5-nitro-benzophenone, hrphzuapqwjpcz-uhfffaoysa
118591
(2-chloro-5-nitrophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)[N+](=O)[O-])Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 78°C to 84°C
Quantity 25 g
Formula Weight 261.66
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-nitrobenzophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'