missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrobenzaldehyde, +99%, ACROS Organics™

Brand:  Acros Organics 404871000

Code : 29

Additional Details : CAS Number : 6361-21-3 Weight : 0.15000kg

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10696611

  • £111.40 / 100g


Estimated Shipment
16-05-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-5-nitrobenzaldehyde
6361-21-3
100.0
99%
ClC6H3(NO2)CHO
MFCD00007293
99+%
98.5
≥98.5%
C7H4ClNO3
185.57

Specifications

2-Chloro-5-nitrobenzaldehyde
6361-21-3
Authentic
Glass bottle
100g
Crystalline Powder
99%
ClC6H3(NO2)CHO
MFCD00007293
99+%
98.5
100.0
77° to 81°C
Pale yellow
≥98.5%
C7H4ClNO3
185.57
Videos
SDS