missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitro-3-picoline, 98%, Thermo Scientific™

Catalog Number p-3736004
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
22280-56-4
C6H5ClN2O2
172.57
MFCD00173686
OSIOIGXJUZTWRI-UHFFFAOYSA-N
2-chloro-5-nitro-3-picoline, pyridine, 2-chloro-3-methyl-5-nitro, 2-chloro-3-methyl-5-nitro pyridine, 2-chloro-3-methyl-5-nitro-pyridine, 2-chloro-5-nitro-3-methylpyridine, pubchem10621, acmc-209fub, ksc207a2n, osioigxjuztwri-uhfffaoysa
89648
2-chloro-3-methyl-5-nitropyridine
CC1=CC(=CN=C1Cl)[N+](=O)[O-]
This item is not returnable. View return policy
22280-56-4
C6H5ClN2O2
172.57
MFCD00173686
OSIOIGXJUZTWRI-UHFFFAOYSA-N
2-chloro-5-nitro-3-picoline, pyridine, 2-chloro-3-methyl-5-nitro, 2-chloro-3-methyl-5-nitro pyridine, 2-chloro-3-methyl-5-nitro-pyridine, 2-chloro-5-nitro-3-methylpyridine, pubchem10621, acmc-209fub, ksc207a2n, osioigxjuztwri-uhfffaoysa
89648
2-chloro-3-methyl-5-nitropyridine
CC1=CC(=CN=C1Cl)[N+](=O)[O-]
CAS Min % 97.5
CAS Max % 100.0
Melting Point 78.0°C to 80.0°C
Color Beige to Cream
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 172.57
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-nitro-3-picoline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-5-nitro-3-picoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'