missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-methylphenol, 99%, Thermo Scientific Chemicals
72.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
2-Chloro-5-methylphenol, is used as a pharmaceutical intermediate. It is also used to make other chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloro-5-methylphenol, is used as a pharmaceutical intermediate. It is also used to make other chemicals.
Solubility
Slightly Soluble (1.2 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
2-Chloro-5-methylphenol, is used as a pharmaceutical intermediate. It is also used to make other chemicals.
Solubility
Slightly Soluble (1.2 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Melting Point | 44°C to 48°C |
Density | 1.215 |
Boiling Point | 195°C to 197°C |
Flash Point | 81°C (177°F) |
Quantity | 100 g |
UN Number | UN3437 |
Beilstein | 2041487 |
Solubility Information | Slightly Soluble (1.2g/L) (25°C). |
Formula Weight | 142.59 |
Percent Purity | 99% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'