missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methylphenol, 99%, Thermo Scientific Chemicals

Catalog Number 11354304
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
615-74-7
C7H7ClO
142.58
MFCD00002175
SMFHPCZZAAMJJO-UHFFFAOYSA-N
6-chloro-m-cresol, phenol, 2-chloro-5-methyl, m-cresol, 6-chloro, 3-methyl-6-chlorophenol, 4-chloro-3-hydroxytoluene, 5-methyl-2-chlorophenol, 6-chloro-3-methylphenol, dsstox_cid_26558, dsstox_rid_81718
12008
2-chloro-5-methylphenol
CC1=CC=C(Cl)C(O)=C1
This item is not returnable. View return policy
615-74-7
C7H7ClO
142.58
MFCD00002175
SMFHPCZZAAMJJO-UHFFFAOYSA-N
6-chloro-m-cresol, phenol, 2-chloro-5-methyl, m-cresol, 6-chloro, 3-methyl-6-chlorophenol, 4-chloro-3-hydroxytoluene, 5-methyl-2-chlorophenol, 6-chloro-3-methylphenol, dsstox_cid_26558, dsstox_rid_81718
12008
2-chloro-5-methylphenol
CC1=CC=C(Cl)C(O)=C1

2-Chloro-5-methylphenol, is used as a pharmaceutical intermediate. It is also used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-5-methylphenol, is used as a pharmaceutical intermediate. It is also used to make other chemicals.

Solubility
Slightly Soluble (1.2 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY
Melting Point 44°C to 48°C
Density 1.215
Boiling Point 195°C to 197°C
Flash Point 81°C (177°F)
Quantity 25 g
UN Number UN3437
Beilstein 2041487
Solubility Information Slightly Soluble (1.2g/L) (25°C).
Formula Weight 142.59
Percent Purity 99%
Chemical Name or Material 2-Chloro-5-methylphenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'